Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HBB2
Entrez Gene ID
100009084
Gene Name
hemoglobin, beta
Gene Aliases
HBB, HBB1
Chromosome Location
Chr.1: 146237016 - 146238517 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082260.3 NP_001075729.2 1 - 1 259 89
NM_001314043.1 NP_001300972.1 1 - 1 259 89
GenBank mRNA M10525.1 - 1 - 1 45 89
M10843.1 - 1 - 1 47 89
V00879.1 - - 47 89

View Full Product Details