Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLPS
Entrez Gene ID
100009061
Gene Name
colipase
Gene Aliases
-
Chromosome Location
Chr.12: 25819529 - 25821125 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082243.1 NP_001075712.1 1 - 2 105 75
GenBank mRNA L06329.1 - 1 - 2 121 75

View Full Product Details