Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB5
Entrez Gene ID
100302410
Gene Name
ankyrin repeat and SOCS box containing 5
Gene Aliases
ASB-5
Chromosome Location
Chr.2: 53701694 - 53776078 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163067.1 NP_001156539.1 2 - 3 387 82
XM_008274050.2 XP_008272272.1 2 - 3 376 82
XM_008274053.2 XP_008272275.1 2 - 3 165 82
XM_008274060.2 XP_008272282.1 2 - 3 278 82
XM_017350167.1 XP_017205656.1 3 - 4 377 82
GenBank mRNA AY165034.1 - 2 - 3 387 82

View Full Product Details