Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPPA5
Entrez Gene ID
100328658
Gene Name
developmental pluripotency associated 5
Gene Aliases
-
Chromosome Location
Chr.12: 60171996 - 60172785 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171255.1 NP_001164726.1 2 - 3 292 71

View Full Product Details