Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP3A6
Entrez Gene ID
100328954
Gene Name
cytochrome P450 3A6
Gene Aliases
-
Chromosome Location
Chr.Unknown: 607005 - 640099 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171268.2 NP_001164739.1 - 1464 76
GenBank mRNA J05034.1 - - 1465 76
M19139.1 - - 1484 76

View Full Product Details