Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCC1
Entrez Gene ID
100328637
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 16713988 - 16717149 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171279.1 NP_001164750.1 1 - 2 1033 58

View Full Product Details