Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPRL3
Entrez Gene ID
100328800
Gene Name
NPR3 like, GATOR1 complex subunit
Gene Aliases
MARE
Chromosome Location
Chr.Unknown: 72255 - 110436 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171413.2 NP_001164884.1 - 232 104

View Full Product Details