Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP298
Entrez Gene ID
100328921
Gene Name
cilia and flagella associated protein 298
Gene Aliases
C14H21orf59, C21orf59, RA_m005_jsm24C07r
Chromosome Location
Chr.14: 162600908 - 162609228 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171500.1 NP_001164971.1 2 - 3 313 90

View Full Product Details