Gene Transcripts

Gene Symbol
Plekha5
Entrez Gene ID
246237
Gene Name
pleckstrin homology domain containing, family A member 5
Gene Aliases
Pepp2, TRS1
Chromosome Location
Chr. 4: 174605468 - 174774692 on Build Rnor_6.0
UniGene ID
Rn.118736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763357.1 XP_008761579.1 11, 12 1405
XM_008763358.1 XP_008761580.1 11, 12 1405
XM_008763359.1 XP_008761581.1 11, 12 1405
XM_008763360.1 XP_008761582.1 11, 12 1405
XM_008763361.1 XP_008761583.1 9, 10 2096
XM_008763362.1 XP_008761584.1 10, 11 1387
XM_008763363.1 XP_008761585.1 4, 5 1233
XM_008763364.1 XP_008761586.1 11, 12 1405
GenBank mRNA AF468695.1 - 9, 10 1154

View Full Product Details