Gene Transcripts

Gene Symbol
Tspy1
Entrez Gene ID
25223
Gene Name
testis specific protein, Y-linked 1
Gene Aliases
Tspy
Chromosome Location
Chr. Y: 151941 - 156208 on Build Rnor_6.0
UniGene ID
Rn.126432
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022923.1 NP_075212.1 5 1064
XM_008773672.1 XP_008771894.1 5 1071
GenBank mRNA AF074880.1 - 5 1064
AJ001377.1 - 5 475

View Full Product Details