Gene Transcripts

Gene Symbol
Slc18a2
Entrez Gene ID
25549
Gene Name
solute carrier family 18 (vesicular monoamine transporter), member 2
Gene Aliases
MNAT, VMAT-2, VMAT2
Chromosome Location
Chr. 1: 280422237 - 280457964 on Build Rnor_6.0
UniGene ID
Rn.9686
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013031.1 NP_037163.1 13 1267
XM_006231664.2 XP_006231726.1 13 2108
XM_008760515.1 XP_008758737.1 12 1467
XM_008760516.1 XP_008758738.1 13 2113
XM_008760517.1 XP_008758739.1 13 2108
XM_008760518.1 XP_008758740.1 12 1467
XM_008760519.1 XP_008758741.1 11 1374
XM_008760520.1 XP_008758742.1 10 1290
GenBank mRNA L00603.1 - 13 1267
M97381.1 - 13 1238

View Full Product Details