Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173323.1 | NP_775445.1 | 2 | 884 | |
GenBank mRNA | U27518.1 | - | 2 | 884 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008770098.1 | XP_008768320.1 | 1 | 284 |