Gene Transcripts

Gene Symbol
Zfp13
Entrez Gene ID
287095
Gene Name
zinc finger protein 13
Gene Aliases
Chromosome Location
Chr. 10: 12908643 - 12916784 on Build Rnor_6.0
UniGene ID
Rn.124720
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105765.1 NP_001099235.1 2, 3 429

View Full Product Details