Gene Transcripts

Gene Symbol
Snrnp25
Entrez Gene ID
287170
Gene Name
small nuclear ribonucleoprotein 25 (U11/U12)
Gene Aliases
RGD1310922
Chromosome Location
Chr. 10: 15685999 - 15689334 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246078.2 XP_006246140.1 5 484
XM_006246079.2 XP_006246141.1 5 526

View Full Product Details