Gene Transcripts

Gene Symbol
Olr5
Entrez Gene ID
292561
Gene Name
olfactory receptor 5
Gene Aliases
ratchr1-65852087-65853025_ORF
Chromosome Location
Chr. 1: 70464948 - 70465856 on Build Rnor_6.0
UniGene ID
Rn.142004
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000112.1 NP_001000112.1 1 279

View Full Product Details