Gene Transcripts

Gene Symbol
Lrrc32
Entrez Gene ID
293135
Gene Name
leucine rich repeat containing 32
Gene Aliases
Garp
Chromosome Location
Chr. 1: 163443973 - 163457426 on Build Rnor_6.0
UniGene ID
Rn.66859
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170434.1 NP_001163905.1 3 1309
XM_006229746.2 XP_006229808.1 3 2159
XM_006229747.2 XP_006229809.1 3 2166

View Full Product Details