Gene Transcripts

Gene Symbol
Ifit1lb
Entrez Gene ID
294090
Gene Name
interferon-induced protein with tetratricopeptide repeats 1-like B
Gene Aliases
Ifit1
Chromosome Location
Chr. 1: 252914684 - 252926426 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760355.1 XP_008758577.1 2 2444
XM_220058.5 XP_220058.2 2 3340

View Full Product Details