Gene Transcripts

Gene Symbol
Nthl1
Entrez Gene ID
29541
Gene Name
nth (endonuclease III)-like 1 (E.coli)
Gene Aliases
Nth1
Chromosome Location
Chr. 10: 13996660 - 14002827 on Build Rnor_6.0
UniGene ID
Rn.14632
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105728.1 NP_001099198.1 5, 6 852

View Full Product Details