Gene Transcripts

Gene Symbol
Pigt
Entrez Gene ID
296360
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class T
Gene Aliases
Chromosome Location
Chr. 3: 160944293 - 160954738 on Build Rnor_6.0
UniGene ID
Rn.40489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106540.2 NP_001100010.2 7 865
XM_006235532.2 XP_006235594.1 7 2151

View Full Product Details