Gene Transcripts

Gene Symbol
Gabrr1
Entrez Gene ID
29694
Gene Name
gamma-aminobutyric acid (GABA) A receptor, rho 1
Gene Aliases
Chromosome Location
Chr. 5: 48345279 - 48411170 on Build Rnor_6.0
UniGene ID
Rn.33552
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017291.3 NP_058987.2 6 747
XM_008763609.1 XP_008761831.1 6 530
GenBank mRNA U21070.1 - 6 704
X95579.1 - 6 747

View Full Product Details