Gene Transcripts

Gene Symbol
Cntnap5c
Entrez Gene ID
297846
Gene Name
contactin associated protein-like 5C
Gene Aliases
Caspr5-4, Cntnap5, Cntnap5d, RGD1566343
Chromosome Location
Chr. 13: 6312877 - 7170811 on Build Rnor_6.0
UniGene ID
Rn.234265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047866.1 NP_001041331.1 674

View Full Product Details