Gene Transcripts

Gene Symbol
Lao1
Entrez Gene ID
298483
Gene Name
L-amino acid oxidase 1
Gene Aliases
Chromosome Location
Chr. 5: 138011760 - 138021653 on Build Rnor_6.0
UniGene ID
Rn.158854
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106682.1 NP_001100152.1 5 617
GenBank mRNA BC100634.1 - 5 617
BC168679.1 - 5 617 5621187

View Full Product Details