Gene Transcripts

Gene Symbol
Dlst
Entrez Gene ID
299201
Gene Name
dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex)
Gene Aliases
Chromosome Location
Chr. 6: 108936549 - 108961322 on Build Rnor_6.0
UniGene ID
Rn.99702
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006981.2 NP_001006982.2 15 1393
XM_006240306.1 XP_006240368.1 13 1425
XM_008764756.1 XP_008762978.1 14 1324
GenBank mRNA BC083858.1 - 15 1379
FQ217447.1 - 14 1318
FQ217539.1 - 14 1318

View Full Product Details