Gene Transcripts

Gene Symbol
Hepacam
Entrez Gene ID
300517
Gene Name
hepatic and glial cell adhesion molecule
Gene Aliases
RGD1306811
Chromosome Location
Chr. 8: 39848255 - 39865424 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729891.4 XP_002729937.2 5 1088

View Full Product Details