Gene Transcripts

Gene Symbol
Atm
Entrez Gene ID
300711
Gene Name
ATM serine/threonine kinase
Gene Aliases
Chromosome Location
Chr. 8: 58015938 - 58119973 on Build Rnor_6.0
UniGene ID
Rn.214048
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106821.1 NP_001100291.1 7, 8 1122
XM_006243068.1 XP_006243130.1 7, 8 1090
XM_008766222.1 XP_008764444.1 8, 9 1183

View Full Product Details