Gene Transcripts

Gene Symbol
Khdc3
Entrez Gene ID
300826
Gene Name
KH domain containing 3, subcortical maternal complex member
Gene Aliases
Ecat1, RGD1311617
Chromosome Location
Chr. 8: 79781941 - 79783845 on Build Rnor_6.0
UniGene ID
Rn.218611
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106837.1 NP_001100307.1 1 79

View Full Product Details