Gene Transcripts

Gene Symbol
Vom2r79
Entrez Gene ID
301137
Gene Name
vomeronasal 2 receptor, 79
Gene Aliases
Chromosome Location
Chr. 9: 9214429 - 9263050 on Build Rnor_6.0
UniGene ID
Rn.217942
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099504.1 NP_001092974.1 3 598

View Full Product Details