Gene Transcripts

Gene Symbol
Mrpl44
Entrez Gene ID
301552
Gene Name
mitochondrial ribosomal protein L44
Gene Aliases
Chromosome Location
Chr. 9: 85542763 - 85547814 on Build Rnor_6.0
UniGene ID
Rn.3298
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031650.1 NP_001026820.1 4 933
GenBank mRNA BC098715.1 - 4 933
FQ215293.1 - 4 1010

View Full Product Details