Gene Transcripts

Gene Symbol
Mageb6
Entrez Gene ID
302441
Gene Name
melanoma antigen family B, 6
Gene Aliases
Mageb17, RGD1565662
Chromosome Location
Chr. X: 60863372 - 60864521 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_228605.6 XP_228605.4 2 977

View Full Product Details