Gene Transcripts

Gene Symbol
Magea8
Entrez Gene ID
302638
Gene Name
melanoma antigen, family A, 8
Gene Aliases
RGD1565778
Chromosome Location
Chr. X: 43172647 - 43184664 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773233.1 XP_008771455.1 4 4422
XM_228890.3 XP_228890.1 5 2890

View Full Product Details