Gene Transcripts

Gene Symbol
Neurl1b
Entrez Gene ID
303019
Gene Name
neuralized E3 ubiquitin protein ligase 1B
Gene Aliases
Neurl2, RGD1564984
Chromosome Location
Chr. 10: 17048251 - 17075139 on Build Rnor_6.0
UniGene ID
Rn.4154
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142652.1 NP_001136124.1 2 455
GenBank mRNA GQ414761.1 - 2 455

View Full Product Details