Gene Transcripts

Gene Symbol
Tbc1d16
Entrez Gene ID
303734
Gene Name
TBC1 domain family, member 16
Gene Aliases
Chromosome Location
Chr. 10: 108257492 - 108327479 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247938.2 XP_006248000.1 4 1251
XM_008768486.1 XP_008766708.1 5 3549
XM_221188.8 XP_221188.5 4 1251

View Full Product Details