Gene Transcripts

Gene Symbol
Sacs
Entrez Gene ID
305940
Gene Name
sacsin molecular chaperone
Gene Aliases
RGD1305416
Chromosome Location
Chr. 15: 41448084 - 41530412 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252167.2 XP_006252229.1 11 11599
XM_006252168.2 XP_006252230.1 10 11127
XM_006252169.2 XP_006252231.1 10 11572
XM_008770792.1 XP_008769014.1 10 11452

View Full Product Details