Gene Transcripts

Gene Symbol
Iglon5
Entrez Gene ID
308557
Gene Name
IgLON family member 5
Gene Aliases
RGD1305344
Chromosome Location
Chr. 1: 98440186 - 98457942 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_218634.8 XP_218634.5 5, 6 634

View Full Product Details