Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006229357.2 | XP_006229419.1 | 6 | 1695 | |
XM_006229358.2 | XP_006229420.1 | 5 | 1489 | ||
XM_008759572.1 | XP_008757794.1 | 6 | 1817 | ||
XM_008759573.1 | XP_008757795.1 | 6 | 1776 | ||
XM_008759574.1 | XP_008757796.1 | 5 | 1872 |