Gene Transcripts

Gene Symbol
Prkrir
Entrez Gene ID
308845
Gene Name
protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor)
Gene Aliases
Chromosome Location
Chr. 1: 163657296 - 163679191 on Build Rnor_6.0
UniGene ID
Rn.161856
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191630.1 NP_001178559.1 5 1761
XM_008759719.1 XP_008757941.1 7 2209

View Full Product Details