Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001044241.1 | NP_001037706.1 | 3, 4 | 661 | |
XM_006230812.2 | XP_006230874.1 | 3, 4 | 852 | ||
GenBank mRNA | DQ450893.1 | - | 3, 4 | 661 | |
FQ221502.1 | - | 3, 4 | 475 | ||
FQ226220.1 | - | 3, 4 | 478 | ||
FQ230464.1 | - | 3, 4 | 729 | ||
FQ230893.1 | - | 3, 4 | 729 |