Gene Transcripts

Gene Symbol
Epb41l2
Entrez Gene ID
309557
Gene Name
erythrocyte membrane protein band 4.1-like 2
Gene Aliases
Epb4.1l2, RGD1563977
Chromosome Location
Chr. 1: 20881820 - 21067713 on Build Rnor_6.0
UniGene ID
Rn.1362
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006227709.1 XP_006227771.1 6 1049
XM_008758646.1 XP_008756868.1 6 1231
XM_008758647.1 XP_008756869.1 6 1347
XM_008758648.1 XP_008756870.1 6 952
XM_008758649.1 XP_008756871.1 7 1187
XM_008758650.1 XP_008756872.1 6 4034
GenBank mRNA FQ212032.1 - 7 1145

View Full Product Details