Gene Transcripts

Gene Symbol
Mdc1
Entrez Gene ID
309595
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
Chromosome Location
Chr. 20: 3405017 - 3419831 on Build Rnor_6.0
UniGene ID
Rn.106312
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166275.1 NP_001159747.1 3 599
GenBank mRNA BC085955.1 - 3 599

View Full Product Details