Gene Transcripts

Gene Symbol
Sobp
Entrez Gene ID
309860
Gene Name
sine oculis-binding protein homolog (Drosophila)
Gene Aliases
RGD1560479, Sobpl, jcx1
Chromosome Location
Chr. 20: 47731713 - 47910442 on Build Rnor_6.0
UniGene ID
Rn.208930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104640.1 NP_001098110.1 6 2379
XM_006256574.2 XP_006256636.1 6 2446
GenBank mRNA DQ503410.1 - 6 2379

View Full Product Details