Gene Transcripts

Gene Symbol
Lrig2
Entrez Gene ID
310753
Gene Name
leucine-rich repeats and immunoglobulin-like domains 2
Gene Aliases
Chromosome Location
Chr. 2: 206928708 - 206997914 on Build Rnor_6.0
UniGene ID
Rn.162624
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107710.1 NP_001101180.1 5 818
XM_006233083.2 XP_006233145.1 5 1213

View Full Product Details