Gene Transcripts

Gene Symbol
Doxl1
Entrez Gene ID
312316
Gene Name
diamine oxidase-like protein 1
Gene Aliases
Chromosome Location
Chr. 4: 78563625 - 78567241 on Build Rnor_6.0
UniGene ID
Rn.79162
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199233.1 NP_954703.1 2 1585

View Full Product Details