Gene Transcripts

Gene Symbol
Utp11l
Entrez Gene ID
313581
Gene Name
UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast)
Gene Aliases
Cgi-94, Cgi94, RGD1559610
Chromosome Location
Chr. 5: 142666288 - 142681173 on Build Rnor_6.0
UniGene ID
Rn.64865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107978.2 NP_001101448.1 5 467
GenBank mRNA BC168964.1 - 5 467
FQ226285.1 - 5 479
FQ231750.1 - 5 488
FQ233930.1 - 5 479

View Full Product Details