Gene Transcripts

Gene Symbol
Tmtc3
Entrez Gene ID
314785
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
RGD1306351
Chromosome Location
Chr. 7: 40171957 - 40217056 on Build Rnor_6.0
UniGene ID
Rn.3901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135858.1 NP_001129330.1 12 1747
XM_008765316.1 XP_008763538.1 11 1657

View Full Product Details