Gene Transcripts

Gene Symbol
Nckap5l
Entrez Gene ID
315297
Gene Name
NCK-associated protein 5-like
Gene Aliases
RGD1561571
Chromosome Location
Chr. 7: 141081943 - 141093924 on Build Rnor_6.0
UniGene ID
Rn.234015
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271369.1 NP_001258298.1 6 1652

View Full Product Details