Gene Transcripts

Gene Symbol
Chordc1
Entrez Gene ID
315447
Gene Name
cysteine and histidine-rich domain (CHORD)-containing 1
Gene Aliases
CHP-1
Chromosome Location
Chr. 8: 17421552 - 17445513 on Build Rnor_6.0
UniGene ID
Rn.6136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108128.1 NP_001101598.1 4 373

View Full Product Details