Gene Transcripts

Gene Symbol
Odf3l1
Entrez Gene ID
315695
Gene Name
outer dense fiber of sperm tails 3-like 1
Gene Aliases
RGD1304976
Chromosome Location
Chr. 8: 61516249 - 61542943 on Build Rnor_6.0
UniGene ID
Rn.76080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108151.1 NP_001101621.1 4 801
XM_006243122.2 XP_006243184.1 5 1767
XM_008766272.1 XP_008764494.1 5 4572

View Full Product Details