Gene Transcripts

Gene Symbol
Prickle4
Entrez Gene ID
316212
Gene Name
prickle homolog 4 (Drosophila)
Gene Aliases
RGD1563979
Chromosome Location
Chr. 9: 15306901 - 15313063 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766897.1 XP_008765119.1 7 1775

View Full Product Details