Gene Transcripts

Gene Symbol
Tcte1
Entrez Gene ID
316242
Gene Name
t-complex-associated testis expressed 1
Gene Aliases
Chromosome Location
Chr. 9: 17847871 - 17867283 on Build Rnor_6.0
UniGene ID
Rn.234588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108206.1 NP_001101676.1 2 516
XM_006244539.2 XP_006244601.1 3 1039
XM_006244540.2 XP_006244602.1 4 1116

View Full Product Details