Gene Transcripts

Gene Symbol
Zcchc5
Entrez Gene ID
317221
Gene Name
zinc finger, CCHC domain containing 5
Gene Aliases
RGD1564805
Chromosome Location
Chr. X: 77953574 - 77957261 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257179.2 XP_006257241.1 2 1773

View Full Product Details